missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 1-EBIO

Description
Activator of epithelial KCa channels, stimulates a large and sustained trans-epithelial Cl- secretory response across T84 monolayers. Induces hyperpolarization to the same magnitude as ACh in aortic value endothelial cells. Promotes embryonic stem cell (ESC) differentiation into cardiomyocytes.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 162.19 |
| Purity | >99% |
| Molecular Weight (g/mol) | 162.19 |
| Chemical Name or Material | 1-EBIO |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?