missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecyne, TRC

Product Code. 30350715
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Product Code. Quantity unitSize
30350715 2.5 g 2.5g
1 options
This item is not returnable. View return policy

Product Code. 30350715

Brand: Toronto Research Chemicals TRCD2627352.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Dodecyne
CAS 765-03-7
Molecular Formula C12H22
Quantity 2.5 g
InChI Formula InChI=1S/C12H22/c1-3-5-7-9-11-12-10-8-6-4-2/h1H,4-12H2,2H3
SMILES CCCCCCCCCCC#C
IUPAC Name dodec-1-yne
Molecular Weight (g/mol) 166.3
Formula Weight 166.1722
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.