missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecylphosphonic acid, 95%

Product Code. 11376729
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376729 1 g 1g
11386729 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376729 Supplier Thermo Scientific Alfa Aesar Supplier No. H26259.03
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5137-70-2 | C12H27O3P | 250.32 g/mol

1-Dodecylphosphonic acid is used as tuning of surface energy, wetting, and work function of metal oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecylphosphonic acid is used as tuning of surface energy, wetting, and work function of metal oxides.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5137-70-2
Molecular Formula C12H27O3P
Molecular Weight (g/mol) 250.32
MDL Number MFCD00015832
InChI Key SVMUEEINWGBIPD-UHFFFAOYSA-N
Synonym n-dodecylphosphonic acid, phosphonic acid, dodecyl, n-dodecanephosphonic acid, dodecane phosphonic acid, 1-dodecanephosphonic acid, n-dodecyl phosphonic acid, dodecyl-phosphonic acid, 1-dodecylphosphonic acid, phosphonic acid, p-dodecyl, n-dodecylphosphonicacid
PubChem CID 78816
IUPAC Name dodecylphosphonic acid
SMILES CCCCCCCCCCCCP(O)(O)=O

Specifications

Melting Point 92°C to 96°C
Quantity 1 g
UN Number UN3261
Solubility Information Sparingly soluble in water.
Formula Weight 250.31
Percent Purity 95%
Chemical Name or Material 1-Dodecylphosphonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.