missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecylamine hydrochloride, 97%

Product Code. 11348735
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348735 10 g 10g
11358735 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348735 Supplier Thermo Scientific Alfa Aesar Supplier No. L12314.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 929-73-7 | C12H28ClN | 221.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-73-7
Molecular Formula C12H28ClN
Molecular Weight (g/mol) 221.81
MDL Number MFCD00042017
InChI Key TWFQJFPTTMIETC-UHFFFAOYSA-N
Synonym dodecylamine hydrochloride, laurylamine hydrochloride, n-dodecylamine hydrochloride, dodecan-1-amine hydrochloride, 1-dodecanamine, hydrochloride, dodecylamine, hydrochloride, unii-1v1470j2mf, 1-dodecanamine, hydrochloride 1:1, armeen 12d hydrochloride, acmc-209rjd
PubChem CID 458426
IUPAC Name dodecan-1-amine;hydrochloride
SMILES [Cl-].CCCCCCCCCCCC[NH3+]

Specifications

Melting Point 186°C to 188°C
Linear Formula CH3(CH2)11NH2·HCl
Quantity 10 g
Beilstein 3553943
Formula Weight 221.82
Percent Purity 97%
Chemical Name or Material 1-Dodecylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.