missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecylamine, 97%

Product Code. 11468750
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 11468750

Brand: Thermo Scientific Alfa Aesar A15515.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-22-1 | C12H27N | 185.36 g/mol

Dodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.

Solubility
Soluble in water (78 mg/L 25°C).

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-22-1
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.36
MDL Number MFCD00008154
InChI Key JRBPAEWTRLWTQC-UHFFFAOYSA-N
Synonym dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4
PubChem CID 13583
IUPAC Name dodecan-1-amine
SMILES CCCCCCCCCCCCN

Specifications

Melting Point 27°C to 31°C
Density 0.802
Boiling Point 64°C to 66°C (0.05 mmHg)
Flash Point 115°C (239°F)
Assay Percent Range ≥98%
Quantity 1 kg
UN Number UN3259
Beilstein 1633576
Solubility Information Soluble in water (78mg/L 25°C).
Formula Weight 185.36
Percent Purity ≥98%
Chemical Name or Material 1-Dodecylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.