missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecylamine, 97%

Product Code. 11462917
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462917 100 g 100g
11472917 500 g 500g
11468750 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462917 Supplier Thermo Scientific Alfa Aesar Supplier No. A15515.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-22-1 | C12H27N | 185.36 g/mol

Dodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.

Solubility
Soluble in water (78 mg/L 25°C).

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-22-1
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.36
MDL Number MFCD00008154
InChI Key JRBPAEWTRLWTQC-UHFFFAOYSA-N
Synonym dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4
PubChem CID 13583
IUPAC Name dodecan-1-amine
SMILES CCCCCCCCCCCCN

Specifications

Melting Point 27°C to 31°C
Density 0.802
Boiling Point 64°C to 66°C (0.05 mm)
Flash Point 115°C (239°F)
Assay Percent Range ≥98%
Quantity 100 g
UN Number UN3259
Beilstein 1633576
Solubility Information Soluble in water (78mg/L 25°C).
Formula Weight 185.36
Percent Purity ≥98%
Chemical Name or Material 1-Dodecylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.