missing translation for 'onlineSavingsMsg'
Learn More

(1-Dodecyl)trimethylammonium bromide, 99%

Product Code. 11489652
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489652 250 g 250g
11469652 10 g 10g
11479652 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489652 Supplier Thermo Scientific Alfa Aesar Supplier No. A10761.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1119-94-4 | C15H34BrN | 308.348 g/mol

(1-Dodecyl)trimethylammonium bromide is used as a surfactant, as an emulsifier of rubber and asphalt and as an anti-static agent of synthetic fiber. It acts as an ionic surfactant and useful in the preparation of gold nanopartilces along with sodium dodecylsulfate. It is also used as paint strippers and foaming stabilizers. It is used to prepare dodecyltrimethylammonium bromide. By using conductimetry, density measurements, and small angle neutron scattering, it is used to study the micelle formation in water-dimethylsulfoxide mixture. Further, it is a useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Dodecyl)trimethylammonium bromide is used as a surfactant, as an emulsifier of rubber and asphalt and as an anti-static agent of synthetic fiber. It acts as an ionic surfactant and useful in the preparation of gold nanopartilces along with sodium dodecylsulfate. It is also used as paint strippers and foaming stabilizers. It is used to prepare dodecyltrimethylammonium bromide. By using conductimetry, density measurements, and small angle neutron scattering, it is used to study the micelle formation in water-dimethylsulfoxide mixture. Further, it is a useful biochemical for proteomics research.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-94-4
Molecular Formula C15H34BrN
Molecular Weight (g/mol) 308.348
MDL Number MFCD00011767
InChI Key XJWSAJYUBXQQDR-UHFFFAOYSA-M
Synonym dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide
PubChem CID 14249
ChEBI CHEBI:282662
IUPAC Name dodecyl(trimethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Br-]

Specifications

Melting Point ∼245°C (decomposition)
Quantity 250 g
UN Number UN3077
Beilstein 3597463
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 308.35
Percent Purity 99%
Chemical Name or Material (1-Dodecyl)trimethylammonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.