missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecene, 96%

Product Code. 11477207
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477207 500 g 500g
11467207 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477207 Supplier Thermo Scientific Alfa Aesar Supplier No. A14992.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 112-41-4 | C12H24 | 168.324 g/mol

1-Dodecene, is used in Oil and Gas field drilling and production operations, in polymer production, as an intermediate, coating, cleaning agent, lubricants and also used as a functional fluid. It is also used in the production of detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecene, is used in Oil and Gas field drilling and production operations, in polymer production, as an intermediate, coating, cleaning agent, lubricants and also used as a functional fluid. It is also used in the production of detergents.

Solubility
It is soluble in ethanol, ethyl ether, and acetone

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-41-4
Molecular Formula C12H24
Molecular Weight (g/mol) 168.324
MDL Number MFCD00008961
InChI Key CRSBERNSMYQZNG-UHFFFAOYSA-N
Synonym 1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr
PubChem CID 8183
IUPAC Name dodec-1-ene
SMILES CCCCCCCCCCC=C

Specifications

Melting Point -37°C
Density 0.758
Boiling Point 212°C to 213°C
Flash Point 73°C (163°F)
Linear Formula CH3(CH2)9CH=CH2
Refractive Index 1.43
Quantity 500 g
Beilstein 1699848
Solubility Information It is soluble in ethanol,ethyl ether,and acetone
Formula Weight 168.32
Percent Purity 96%
Chemical Name or Material 1-Dodecene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Dodecene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: 1-Dodecene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.