missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecene, 93-95%

Product Code. p-124133 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Metal Drum
Plastic drum
Unit Size:
100g
10kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10071070 100 g Glass bottle 100g
10449410 500 g Glass bottle 500g
10108043 2.5 kg Plastic drum 2.5kg
11964421 10 kg Metal Drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10071070 Supplier Thermo Scientific Acros Supplier No. 117631000
Add to basket
This item is not returnable. View return policy

CAS: 112-41-4 | C12H24 | 168.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-41-4
Molecular Formula C12H24
Molecular Weight (g/mol) 168.32
MDL Number MFCD00008961
InChI Key CRSBERNSMYQZNG-UHFFFAOYSA-N
Synonym 1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr
PubChem CID 8183
IUPAC Name dodec-1-ene
SMILES CCCCCCCCCCC=C

Specifications

CAS Min % 93.0
CAS Max % 100.0
Melting Point -35.0°C
Density 0.7500g/mL
Boiling Point 213.0°C
Flash Point 77°C
Assay Percent Range 93% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)9CH=CH2
Refractive Index 1.4284 to 1.4304
Quantity 100 g
Beilstein 01,225
Specific Gravity 0.75
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether
Formula Weight 168.32
Percent Purity 93 to 95%
Chemical Name or Material 1-Dodecene, 93-95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.