missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanol, TRC

Product Code. 17982405
Change view
Click to view available options
Quantity:
5 g
500 g
Unit Size:
500g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982405 5 g 5g
30132507 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17982405 Supplier Toronto Research Chemicals Supplier No. TRCD5254605G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-53-8
Molecular Formula C12 H26 O
Molecular Weight (g/mol) 186.33
Synonym 1-Dodecanol (9CI, ACI), Dodecyl alcohol (8CI), 1-Dodecyl alcohol, 1-Hydroxydodecane, Adol 10, Adol 11, Adol 12, Alfol 12, C 1214N, Conol 1275, Conol 20P, Conol 20PP, Epal 12, Hainol 12SS, K 20, K 20 (alcohol), Kalcohl 20, Kalcohl 2098, Kalcol 2098, Lauric alcohol, Laurinic alcohol, Lauryl 24, Lauryl alcohol, Lipocol L, Lorol C 12, Lorol C 12/98, n-Dodecan-1-ol, n-Dodecyl alcohol, NAA 42, Nacol 12-96, Nacol 12-99, NSC 3724, Pisol, S 1298, Sipol L 12, Siponol 25, Siponol L 2, Siponol L 5
IUPAC Name dodecan-1-ol
SMILES CCCCCCCCCCCCO

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H26O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3
Formula Weight 186.1984
Chemical Name or Material 1-Dodecanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.