missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanol, ACS Reagent, ≥98.0%, Honeywell Riedel-de Haen™

Product Code. 16683954
Change view
Click to view available options
Quantity:
5 L
25 kg
Unit Size:
25kg
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16683954 5 L 5 L
16673954 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16683954 Supplier Honeywell Riedel-de Haën Supplier No. 4438165L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 112-53-8
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
InChI Key LQZZUXJYWNFBMV-UHFFFAOYSA-N
IUPAC Name dodecan-1-ol
SMILES CCCCCCCCCCCCO

Specifications

CAS Max % ≤ 100%
Melting Point 22°C to 26°C
Color White
Boiling Point 260°C to 262°C
Flash Point 121°C
Quantity 5 L
Chemical Name or Material 1-Dodecanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.