missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Dodecanol, ACS Reagent, ≥98.0%, Honeywell Riedel-de Haen™
Click to view available options
Quantity:
5 L
25 kg
Unit Size:
25kg
5 L
Specifications
Specifications
| CAS Max % | ≤ 100% |
| Melting Point | 22°C to 26°C |
| Color | White |
| Boiling Point | 260°C to 262°C |
| Flash Point | 121°C |
| Linear Formula | CH3(CH2)11OH |
| Quantity | 25 kg |
| Beilstein | 1738860 |
| Grade | ACS General Use Solvents |
| Chemical Name or Material | 1-Dodecanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction