missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanol, 98%

Product Code. 11424913
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424913 1000 g 1000g
11434913 250 g 250g
11444913 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424913 Supplier Thermo Scientific Alfa Aesar Supplier No. A12228.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-53-8 | C12H26O | 186.34 g/mol

1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.

Solubility
Soluble in water (0.004g/L).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-53-8
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
MDL Number MFCD00004753
InChI Key LQZZUXJYWNFBMV-UHFFFAOYSA-N
Synonym 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol
PubChem CID 8193
ChEBI CHEBI:28878
IUPAC Name dodecan-1-ol
SMILES CCCCCCCCCCCCO

Specifications

Melting Point 24°C to 27°C
Density 0.83
Boiling Point 258°C to 260°C
Flash Point 127°C (260°F)
Assay Percent Range 98%
Odor Alcohol-like
Linear Formula CH3(CH2)11OH
Refractive Index 1.443
Quantity 1000 g
UN Number UN3077
Beilstein 1738860
Merck Index 14,3405
Solubility Information Soluble in water (0.004g/L).
Formula Weight 186.34
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-Dodecanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Dodecanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-Dodecanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.