missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanethiol, 98%

Product Code. 11450703
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450703 500 g 500g
11440703 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11450703 Supplier Thermo Scientific Alfa Aesar Supplier No. A12741.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 112-55-0 | C12H26S | 202.40 g/mol

1-Dodecanethiol is used in the preparation of hydrophobic or mixed self-assembled monolayers. It is also employed as a chain transfer agent for radical polymerization. Further, it is utilized as a polymerization inhibitor in polyurethane and neoprene adhesives, which finds application in the footwear industry. In addition to this, it acts as a protein regenerating agent used for the regeneration of native proteins from mercuribenzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecanethiol is used in the preparation of hydrophobic or mixed self-assembled monolayers. It is also employed as a chain transfer agent for radical polymerization. Further, it is utilized as a polymerization inhibitor in polyurethane and neoprene adhesives, which finds application in the footwear industry. In addition to this, it acts as a protein regenerating agent used for the regeneration of native proteins from mercuribenzoate.

Solubility
Immiscible with water.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-55-0
Molecular Formula C12H26S
Molecular Weight (g/mol) 202.40
MDL Number MFCD00004885
InChI Key WNAHIZMDSQCWRP-UHFFFAOYSA-N
Synonym 1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan
PubChem CID 8195
IUPAC Name dodecane-1-thiol
SMILES CCCCCCCCCCCCS

Specifications

Melting Point -7°C
Density 0.845
Boiling Point 276°C to 278°C
Flash Point 133°C (271°F)
Assay Percent Range 98%
Odor Unpleasant
Linear Formula CH3(CH2)11SH
Refractive Index 1.4601
Quantity 500 g
Beilstein 969337
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 202.4
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Dodecanethiol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Dodecanethiol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin sensitiser Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1-Dodecanethiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.