missing translation for 'onlineSavingsMsg'
Learn More

1-Docosanol, TRC

Product Code. 30384551
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Product Code. Quantity
30384551 10 g
30377056 1 g
30372005 50 g
3 options
This item is not returnable. View return policy

Product Code. 30384551

Brand: Toronto Research Chemicals TRCD67885510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 661-19-8
Molecular Formula C22 H46 O
Molecular Weight (g/mol) 326.6
Synonym 1-Docosanol, 1-Docosonol, Abreva, Behenic alcohol, Behenyl 80 Alcohol, Behenyl alc. 80, Behenyl alcohol, Conol 2265, Conol 2280, Docosanol, Docosyl alcohol, Hainol 22S, IK 2, IK 2 (alcohol), Kalcohl 220, Kalcohl 22080, Kalcol 22080, Lanette 22, NAA 422, NSC 8407, Nacol 22-97, Nacol 22-98, Nacol C 22, Stenol 1822, Stenol 1822A, Tadenan, Toho BH 65, n-Docosanol
IUPAC Name docosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCO

Specifications

Quantity 10 g
InChI Formula InChI=1S/C22H46O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23/h23H,2-22H2,1H3
Formula Weight 326.3549
Chemical Name or Material n-Docosanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.