missing translation for 'onlineSavingsMsg'
Learn More

1-Docosanol-d45, TRC

Product Code. 30381532
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381532 50 mg
30379186 500 mg
30371012 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30381532 Supplier Toronto Research Chemicals Supplier No. TRCD67885750MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C22HD45O
Molecular Weight (g/mol) 371.88
Synonym 1-Docosonol-d45, Abreva-d45, Behenic-d45 Alcohol, Behenyl-d45 80 Alcohol, Behenyl-d45 Alc. 80, Behenyl-d45 Alcohol, Conol-d45 2265, Conol-d45 2280, Docosanol-d45, Docosyl-d45 Alcohol, Hainol-d45 22S, IK-d45 2, IK 2-d45 (alcohol), Kalcohl-d45 220, Kalcohl-d45 22080, Kalcol-d45 22080, Lanette-d45 22, NAA-d45 422, NSC-d45 8407, Nacol-d45 22-97, Nacol-d45 22-98, Nacol-d45 C 22, Stenol-d45 1822, Stenol-d45 1822A, Tadenan-d45, Toho-d45 BH 65, n-Docosanol-d45
SMILES OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H46O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23/h23H,2-22H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2
Chemical Name or Material 1-Docosanol-d45
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.