missing translation for 'onlineSavingsMsg'
Learn More

1-Docosanethiol, TRC

Product Code. 30371245
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30371245 250 mg -
30379508 500 mg -
30378113 100 mg 100mg
3 options
This item is not returnable. View return policy

Product Code. 30371245

Brand: Toronto Research Chemicals TRCD678868250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7773-83-3
Formula Weight 342.332
InChI Formula InChI=1S/C22H46S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23/h23H,2-22H2,1H3
IUPAC Name docosane-1-thiol
Molecular Formula C22H46S
Molecular Weight (g/mol) 342.67
SMILES CCCCCCCCCCCCCCCCCCCCCCS
Synonym Docosanethiol, Docosyl Mercaptan, NSC 97595, n-Docosanethiol
Chemical Name or Material 1-Docosanethiol

Specifications

Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.