missing translation for 'onlineSavingsMsg'
Learn More

1-(Diphenylmethyl)piperazine, 97%, Thermo Scientific Chemicals

Catalog Number 10427510
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
841-77-0
MFCD00038379
NWVNXDKZIQLBNM-UHFFFAOYSA-N
1-diphenylmethyl piperazine, benzhydrylpiperazine, n-benzhydrylpiperazine, 1-benzhydryl-piperazine, diphenylmethylpiperazine, norcyclizine, normethylcyclizine, piperazine, 1-diphenylmethyl, 4-benzhydrylpiperazine, n-diphenylmethyl piperazine
70048
1-benzhydrylpiperazine
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3
This item is not returnable. View return policy
841-77-0
MFCD00038379
NWVNXDKZIQLBNM-UHFFFAOYSA-N
1-diphenylmethyl piperazine, benzhydrylpiperazine, n-benzhydrylpiperazine, 1-benzhydryl-piperazine, diphenylmethylpiperazine, norcyclizine, normethylcyclizine, piperazine, 1-diphenylmethyl, 4-benzhydrylpiperazine, n-diphenylmethyl piperazine
70048
1-benzhydrylpiperazine
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 88°C to 93°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Molecular Formula C17H20N2
Quantity 100 g
Molecular Weight (g/mol) 252.361
Formula Weight 252.36
Percent Purity ≥96.0% Total Base (on dry substance), ≥98.0% (TLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Diphenylmethyl)piperazine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1-(Diphenylmethyl)piperazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'