missing translation for 'onlineSavingsMsg'
Learn More

1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt, 95%, Thermo Scientific™

Product Code. 11489637
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489637 50 g 50g
11479637 10 g 10g
11499637 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489637 Supplier Thermo Scientific Alfa Aesar Supplier No. B22124.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136997-71-2
Molecular Formula C10H14N2O4S
Molecular Weight (g/mol) 258.29
MDL Number MFCD00185917
InChI Key SJHHXGGXZKPIDE-UHFFFAOYSA-N
Synonym 1-n,n-dimethylcarbamoyl-4-2-sulfoethyl pyridinium hydroxide inner salt, 1-dimethylcarbamoyl-4-2-sulfoethyl pyridinium inner salt, 2-1-dimethylcarbamoyl pyridin-1-ium-4-yl ethanesulfonate, 1-dimethylcarbamoyl-4-2-sulfoethyl pyridinium hydroxide inner salt, 1-dimethylcarbamoyl-4-2-sulfonatoethyl pyridin-1-ium, 2-1-n,n-dimethylcarbamoyl-4-pyridyl ethanesulfonic acid, 1-dimethylcarbamoyl-4-2-sulfoethyl pyridinium betaine, 1-dimethylcarbamoyl-4-2-sulfonatoethyl pyridinium, 2-1-dimethylcarbamoyl-4-pyridinio ethylsulfonate
PubChem CID 3853422
SMILES CN(C)C(=O)[N+]1=CC=C(CCS([O-])(=O)=O)C=C1

Specifications

Melting Point ∼180°C (decomposition)
Quantity 50 g
IUPAC Name 1-(dimethylcarbamoyl)-4-(2-sulfonatoethyl)pyridin-1-ium
Formula Weight 258.29
Percent Purity 95%
Chemical Name or Material 1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.