missing translation for 'onlineSavingsMsg'
Learn More

1-Dimethylamino-but-1-en-3-one, 98%

Product Code. 10217560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
10mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10217560 100 mL Glass bottle 100mL
10628401 10 mL Glass bottle 10mL
10733914 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10217560 Supplier Thermo Scientific Acros Supplier No. 298021000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2802-08-6 | C6H11NO | 113.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2802-08-6
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00142611
InChI Key QPWSKIGAQZAJKS-SNAWJCMRSA-N
Synonym e-4-dimethylamino but-3-en-2-one, trans-4-dimethylamino-3-buten-2-one, 4-dimethylamino but-3-en-2-one, 3e-4-dimethylamino but-3-en-2-one, 1-dimethylamino-but-1-en-3-one, 4-dimethylamino-3-buten-2-one, 3e-4-dimethylamino-3-buten-2-one, 3-buten-2-one, 4-dimethylamino-, 3e, 3-buten-2-one, 4-dimethylamino, 3-buten-2-one, 4-dimethylamino-, 3e-9ci
PubChem CID 5369152
IUPAC Name (E)-4-(dimethylamino)but-3-en-2-one
SMILES CC(=O)C=CN(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown to Brown-Red
Density 0.9600g/mL
Boiling Point 101°C to 102°C (5.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCHCHC(O)CH3
Refractive Index 1.5570 to 1.56
Quantity 100 mL
Specific Gravity 0.96
Solubility Information Solubility in water: miscible. Other solubilities: miscible with methanol
Formula Weight 113.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Dimethylamino-but-1-en-3-one
Show More Show Less
compliance-icons
Product Identifier
  • 1-Dimethylamino-but-1-en-3-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.