missing translation for 'onlineSavingsMsg'
Learn More

1-Dimethylamino-2-propanol, 99%

Product Code. p-115306 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10713651 250 g Glass bottle 250g
10366550 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10713651 Supplier Thermo Scientific Acros Supplier No. 115872500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-16-7 | C5H13NO | 103.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-16-7
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.16
MDL Number MFCD00004532
InChI Key NCXUNZWLEYGQAH-UHFFFAOYSA-N
Synonym 1-dimethylamino-2-propanol, dimepranol, 1-dimethylamino propan-2-ol, n,n-dimethylisopropanolamine, dimethyl 2-hydroxypropyl amine, 2-propanol, 1-dimethylamino, dimethylisopropanolamine, dimepranol inn, n,n-dimethylamino-2-propanol
PubChem CID 37511
IUPAC Name 1-(dimethylamino)propan-2-ol
SMILES CC(CN(C)C)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -85.0°C
Color Colorless to Yellow
Density 0.8500g/mL
Boiling Point 126.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCH2CH(OH)CH3
Refractive Index 1.4183 to 1.4203
Quantity 250 g
Specific Gravity 0.85
Solubility Information Solubility in water: miscible with water. Other solubilities: soluble in most organic solvents
Viscosity 4.9 mPa.s (50°C)
Formula Weight 103.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Dimethylamino-2-propanol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Dimethylamino-2-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.