missing translation for 'onlineSavingsMsg'
Learn More

1-Deoxynojirimycin hydrochloride, 98%

Product Code. 15427829
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427829 25 mg 25mg
15417829 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427829 Supplier Thermo Scientific Alfa Aesar Supplier No. J66458.MB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 73285-50-4 | C6H14ClNO4 | 199.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73285-50-4
Molecular Formula C6H14ClNO4
Molecular Weight (g/mol) 199.631
MDL Number MFCD00133247
InChI Key ZJIHMALTJRDNQI-VFQQELCFSA-N
Synonym duvoglustat hcl, duvoglustat hydrochloride, deoxynojirimycin hydrochloride, 1-deoxynojirimycin hydrochloride, unii-0rn23c42qr, duvoglustat hydrochloride usan, +-1-deoxynojirimycin hydrochloride, moranoline hydrochloride, 1-deoxy-l-altronojirimycin hydrochloride
PubChem CID 13018787
IUPAC Name (2R,3R,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
SMILES C1C(C(C(C(N1)CO)O)O)O.Cl

Specifications

Color White
Quantity 25 mg
Beilstein 3563225
Solubility Information Soluble in water at 20mg/ml
Formula Weight 199.64
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Deoxynojirimycin hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.