missing translation for 'onlineSavingsMsg'
Learn More

1-Decyne, 98%, Thermo Scientific Chemicals

Product Code. 10402421
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 764-93-2
Molecular Formula C10H18
Molecular Weight (g/mol) 138.25
MDL Number MFCD00009576
InChI Key ILLHQJIJCRNRCJ-UHFFFAOYSA-N
Synonym 1-decyne, decyne, octylacetylene, unii-ulr28gd98q, ulr28gd98q, decine, n-octylacetylene, deca-1-yne, acmc-209p3y
PubChem CID 12997
ChEBI CHEBI:87322
IUPAC Name dec-1-yne
SMILES CCCCCCCCC#C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -44.0°C
Color Colorless to Yellow
Density 0.7600g/mL
Boiling Point 174.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7C≡CH
Refractive Index 1.4260 to 1.4280
Quantity 25 mL
Beilstein 01,I,122
Specific Gravity 0.76
Formula Weight 138.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Decyne, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Decyne
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'