Learn More
1-Decene, 96%, remainder isomers
Description
1-Decene is the only isomer of industrial importance. it is used as a monomer in copolymers and is an intermediate in the production of epoxides, amines, oxo alcohols, synthetic lubricants, synthetic fatty acids, and alkylated aromatics. 1-Decene has been isolated from the leaves and rhizome of the plant Farfugium japonicum and has been detected as the initial product in the microbial degradation of n-decane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -66°C |
| Color | Colorless |
| Density | 0.741 g/mL |
| Boiling Point | 170°C to 171°C |
| Flash Point | 49°C (120°F) |
| Assay Percent Range | 96% |
| Linear Formula | CH3(CH2)7CH=CH2 |
| Refractive Index | 1.421 |
| Quantity | 500 g |
| UN Number | UN3295 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.