Learn More
Ammonium hexachlororhodate(III) hydrate, Premion™, 99.995% (metals basis)
Description
It is used as an intermediate in organic synthesis for the preparation of pharmaceuticals and agrochemicals. It can be used as a precursor for the functionalization of multi-walled carbon nanotubes (MWCNT) with metallic nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Color | Red |
| Physical Form | Powder |
| Quantity | 0.5 g |
| Solubility Information | Soluble in water |
| IUPAC Name | triazanium;hexachlororhodium(3-) |
| Formula Weight | 369.74 (Anhydrous) |
| Percent Purity | 99.995% |
| Grade | Premion |
| Sensitivity | Hygroscopic |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.