missing translation for 'onlineSavingsMsg'
Learn More

1-Decanoyl-rac-glycerol, TRC

Product Code. 30134271
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134271 2.5 g 2.5g
30003976 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134271 Supplier Toronto Research Chemicals Supplier No. TRCD2126502.5G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2277-23-8
Molecular Formula C13H26O4
Molecular Weight (g/mol) 246.34
Synonym Decanoic Acid 2,3-Dihydroxypropyl Ester, (±)-1-Monodecanoin, (±)-Glyceryl 1-Monodecanoate, 1-Decanoylglycerol, 1-Monocaprin, 1-Monodecanoylglycerol, 2,3-Dihydroxypropyl Decanoate, Capric Acid alpha-Monoglyceride, Capryl Monoglyceride, Decanoic Acid 1-Monoglyceride, Decanoic Acid alpha-Monoglyceride, Glycerol 1-Monodecanoate, alpha-Monocaprin;
IUPAC Name 2,3-dihydroxypropyl decanoate
SMILES CCCCCCCCCC(=O)OCC(O)CO

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C13H26O4/c1-2-3-4-5-6-7-8-9-13(16)17-11-12(15)10-14/h12,14-15H,2-11H2,1H3
Formula Weight 246.1831
Chemical Name or Material 1-Decanoyl-rac-glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.