missing translation for 'onlineSavingsMsg'
Learn More

1-Decanethiol, 96%

Product Code. 11414180
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414180 25 mL 25mL
11428446 100 mL 100mL
11438446 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414180 Supplier Thermo Scientific Alfa Aesar Supplier No. A15913.AC
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 143-10-2 | C10H22S | 174.346 g/mol

1-Decanethiol is used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for field-effect transistors (FETs). It is also used in the study of self-assembled monolayers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decanethiol is used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for field-effect transistors (FETs). It is also used in the study of self-assembled monolayers.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. It is sensitive to air. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-10-2
Molecular Formula C10H22S
Molecular Weight (g/mol) 174.346
MDL Number MFCD00004884
InChI Key VTXVGVNLYGSIAR-UHFFFAOYSA-N
Synonym 1-decanethiol, decyl mercaptan, 1-mercaptodecane, decylmercaptan, decanethiol, decanethiol-1, decanthiol, n-decanethiol, n-decanthiol, 1-decylthiol
PubChem CID 8917
IUPAC Name decane-1-thiol
SMILES CCCCCCCCCCS

Specifications

Melting Point -26°C
Density 0.843
Boiling Point 237°C
Flash Point 98°C (208°F)
Odor Unpleasant
Linear Formula CH3(CH2)9SH
Refractive Index 1.4565
Quantity 25 mL
Beilstein 1735223
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 174.35
Percent Purity 96%
Chemical Name or Material 1-Decanethiol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Decanethiol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1-Decanethiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.