missing translation for 'onlineSavingsMsg'
Learn More

1-Decanesulfonic Acid Sodium Salt, Ion Pair Chromatography, with Electrochemical Detection, Fisher Chemical

Product Code. 10711974 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10711974 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10711974 Supplier Fisher Chemical Supplier No. D/0035/44

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 300°C
CAS 13419-61-9
Color White
Physical Form Solid
Quantity 5 g
Molecular Formula C10H21NaO3S
MDL Number 7526
InChI Key AIMUHNZKNFEZSN-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCS([O-])(=O)=O
IUPAC Name sodium decane-1-sulfonate
Molecular Weight (g/mol) 244.32
Formula Weight 244.32g/mol
Packaging Glass Jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.