missing translation for 'onlineSavingsMsg'
Learn More

1-(Cyclopropylcarbonyl)piperazine hydrochloride, 97%

Product Code. 15430097
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430097 5 g 5g
15440097 250 mg 250mg
15420097 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430097 Supplier Thermo Scientific Alfa Aesar Supplier No. H66242.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1021298-67-8 | C8H15ClN2O | 190.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1021298-67-8
Molecular Formula C8H15ClN2O
Molecular Weight (g/mol) 190.67
MDL Number MFCD03428576
InChI Key WIHDAPMHNYYTOA-UHFFFAOYSA-N
Synonym 1-cyclopropylcarbonyl piperazine hydrochloride, cyclopropyl piperazin-1-yl methanone hydrochloride, 1-cyclopropanecarbonylpiperazine hydrochloride, cyclopropyl piperazin-1-yl methanone hcl, cyclopropylcarboxylic acid-1-piperazineamide hydrochloride, cyclopropyl piperazinyl ketone, chloride, pubchem19546, 1-cyclopropylcarbonyl piperazine hcl, 1-cyclopropylcarbonyl-piperazine hcl, 1-cyclopropylcarbonyl piperazine hcl salt
PubChem CID 20847955
IUPAC Name cyclopropyl(piperazin-1-yl)methanone;hydrochloride
SMILES Cl.O=C(C1CC1)N1CCNCC1

Specifications

Melting Point 175°C to 179°C
Color White
Quantity 5 g
Beilstein 18312533
Formula Weight 190.67
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(Cyclopropylcarbonyl)piperazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.