missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic Acid, Mikromol™

Product Code. 30049704
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30049704 100 mg 100mg
30049168 25 mg 25mg
2 options
This item is not returnable. View return policy

Product Code. 30049704

Brand: Mikromol MM0885.13

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Cyclopropyl-6,7-difluoro-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
CAS 112811-72-0
Analyte or Component Names 1-Cyclopropyl-6,7-difluoro-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Shipping Condition Room Temperature
Molecular Formula C14 H11 F2 N O4
Certifications/Compliance ISO 17025
Synonym 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic Acid, 1-Cyclopropyl-1,4-dihydro-6,7-difluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid, 1-Cyclopropyl-6,7-difluoro-8-methoxy-1,4-dihydro-4-oxoquinoline-3-carboxylic acid, 1-Cyclopropyl-6,7-difluoro-8-methoxy-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid, 1-Cyclopropyl-6,7-difluoro-8-methoxy-4-oxo-1,4-dihydroquinolin-3-carboxylic acid, 1-Cyclopropyl-6,7-difluoro-8-methoxy-4-quinolone-3-carboxylic acid
InChI Formula InChI=1S/C14H11F2NO4/c1-21-13-10(16)9(15)4-7-11(13)17(6-2-3-6)5-8(12(7)18)14(19)20/h4-6H,2-3H2,1H3,(H,19,20)
SMILES COc1c(F)c(F)cc2C(=O)C(=CN(C3CC3)c12)C(=O)O
IUPAC Name 1-cyclopropyl-6,7-difluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid
Molecular Weight (g/mol) 295.24
Quantity 100 mg
Formula Weight 295.0656 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.