missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-hydroxy-4-oxo-3-quinolinecarboxylic Acid, Mikromol™
Description
1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-hydroxy-4-oxo-3-quinolinecarboxylic Acid
Specifications
Specifications
| Chemical Name or Material | 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-hydroxy-4-oxo-3-quinolinecarboxylic Acid |
| CAS | 154093-72-8 |
| Analyte or Component Names | 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-hydroxy-4-oxo-3-quinolinecarboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H9 F2 N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Diketopiperazine, 3-Quinolinecarboxylic acid, 1-cyclopropyl-6,7-difluoro-1,4-dihydro-8-hydroxy-4-oxo-, 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-hydroxy-4-oxo-3-quinolinecarboxylic acid, 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-hydroxy-4-oxo-3-quinolinecarboxylic Acid (8-Hydroxyquinolonic Acid) |
| SMILES | OC(=O)C1=CN(C2CC2)c3c(O)c(F)c(F)cc3C1=O |
| Molecular Weight (g/mol) | 281.21 |
| Quantity | 25 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?