missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexylethanone, TRC

Product Code. 30363409
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363409 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30363409 Supplier Toronto Research Chemicals Supplier No. TRCC9921055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Cyclohexylethanone
CAS 823-76-7
Molecular Formula C8H14O
Quantity 5 g
Synonym 1-Acetylcyclohexane, 1-Cyclohexylacetaldehyde, 1-Cyclohexylethan-1-one, Hexahydroacetophenone, Acetylcyclohexane, Cyclohexyl Methyl Ketone, Cyclohexylethanone, Methyl Cyclohexyl Ketone, NSC 16249
InChI Formula InChI=1S/C8H14O/c1-7(9)8-5-3-2-4-6-8/h8H,2-6H2,1H3
SMILES O=C(C)C1CCCCC1
IUPAC Name 1-cyclohexylethanone
Molecular Weight (g/mol) 126.2
Formula Weight 126.1045
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.