missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexylethanol, 98%

Product Code. 11454267
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454267 25 g 25g
11444267 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454267 Supplier Thermo Scientific Alfa Aesar Supplier No. B20363.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1193-81-3 | C8H16O | 128.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-81-3
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00001475
InChI Key JMSUNAQVHOHLMX-UHFFFAOYSA-N
Synonym methylcyclohexylcarbinol, 1-cyclohexyl-1-ethanol, cyclohexylmethylcarbinol, 1-cyclohexylethan-1-ol, ethanol, 1-cyclohexyl, methanol, cyclohexylmethyl, cyclohexanemethanol, .alpha.-methyl, +--1-cyclohexylethanol, 1-cyclohexyl-ethanol, acmc-20aovg
PubChem CID 137829
IUPAC Name 1-cyclohexylethanol
SMILES CC(C1CCCCC1)O

Specifications

Density 0.92
Boiling Point 188°C to 190°C
Flash Point 73°C (163°F)
Refractive Index 1.465
Quantity 25 g
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 1-Cyclohexylethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.