missing translation for 'onlineSavingsMsg'
Learn More

1-(Cyclohexylcarbonyl)piperazine, 97%, Thermo Scientific™

Product Code. 15431748
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27561-62-2
Molecular Formula C11H20N2O
Molecular Weight (g/mol) 196.294
MDL Number MFCD06798113
InChI Key ZSZROXCAFYZNHE-UHFFFAOYSA-N
Synonym 1-cyclohexylcarbonyl piperazine, cyclohexyl piperazin-1-yl methanone, 1-cyclohexanecarbonylpiperazine, methanone, cyclohexyl-1-piperazinyl, maybridge1_006754, cyclohexylpiperazino ketone, cyclohexyl piperazinyl ketone, n-cyclohexylcarbonyl piperazine, cyclohexyl-1-piperazinylmethanone, 1-cyclohexanecarbonyl-piperazine
PubChem CID 3437502
IUPAC Name cyclohexyl(piperazin-1-yl)methanone
SMILES C1CCC(CC1)C(=O)N2CCNCC2

Specifications

Melting Point 84°C to 89°C
Color White to Brown
Quantity 1 g
Formula Weight 196.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(Cyclohexylcarbonyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'