missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-2-pyrrolidinone, 99%

Product Code. 11437977
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11437977

Brand: Thermo Scientific Alfa Aesar A16638.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6837-24-7 | C10H17NO | 167.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6837-24-7
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00003191
InChI Key PZYDAVFRVJXFHS-UHFFFAOYSA-N
Synonym n-cyclohexyl-2-pyrrolidone, 1-cyclohexyl-2-pyrrolidone, 2-pyrrolidinone, 1-cyclohexyl, cyclohexyl pyrrolidone, n-cyclohexylpyrrolidone, 1-cyclohexyl-2-pyrrolidinone, n-cyclohexylpyrrolidinone, unii-7im8p1vvnm, 7im8p1vvnm, acmc-20aj1a
PubChem CID 81278
IUPAC Name 1-cyclohexylpyrrolidin-2-one
SMILES C1CCC(CC1)N2CCCC2=O

Specifications

Melting Point 12°C to 15°C
Density 1.007
Boiling Point 290°C to 292°C
Flash Point 140°C (284°F)
Refractive Index 1.499
Quantity 50 g
UN Number UN2810
Beilstein 121832
Formula Weight 167.25
Percent Purity 99%
Chemical Name or Material 1-Cyclohexyl-2-pyrrolidinone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.