missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Cyclohexyl-1-propanol, 97%

Product Code. 11474497
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474497 25 g 25g
11464497 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474497 Supplier Alfa Aesar™ Supplier No. B20497.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17264-02-7
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00001476
InChI Key JVTXOMXEPFDMHB-UHFFFAOYNA-N
Synonym 1-cyclohexyl-1-propanol, 1-cyclohexylpropanol, ethylcyclohexylcarbinol, cyclohexanemethanol, .alpha.-ethyl, alpha-ethylcyclohexane-1-methanol, cyclohexanemethanol, alpha-ethyl, acmc-1bowk, 3-cyclohexyl-propanol, 1-cyclohexylpropanol-1, alpha-ethylcyclohexanemethanol, +/-
PubChem CID 86535
IUPAC Name 1-cyclohexylpropan-1-ol
SMILES CCC(O)C1CCCCC1

Specifications

Density 0.9
Flash Point 84°C (183°F)
Quantity 25 g
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material 1-Cyclohexyl-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.