missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Cyclohexenylcarboxylic acid, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 1-Cyclohexenylcarboxylic acid |
| CAS | 636-82-8 |
| Molecular Formula | C7H10O2 |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C7H10O2/c8-7(9)6-4-2-1-3-5-6/h4H,1-3,5H2,(H,8,9) |
| SMILES | OC(=O)C1=CCCCC1 |
| IUPAC Name | cyclohexene-1-carboxylic acid |
| Molecular Weight (g/mol) | 126.15 |
| Formula Weight | 126.0681 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction