missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexene-1-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 11439622
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.

Solubility
Soluble in water (0.7 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-82-8
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00001545
InChI Key NMEZJSDUZQOPFE-UHFFFAOYSA-N
Synonym 1-cyclohexene-1-carboxylic acid, 1-cyclohexenecarboxylic acid, cyclohex-1-enecarboxylic acid, cyclohexenecarboxylic acid, cyclohex-1-ene-1-carboxylic acid, 1-cyclohexenecarboxylicacid, cyclohexenyl carboxylic acid, 1-cyclohexene carboxylic acid, acmc-1b731, 3,4,5,6-tetrahydrobenzoic acid
PubChem CID 69470
ChEBI CHEBI:29565
IUPAC Name cyclohexene-1-carboxylic acid
SMILES C1CCC(=CC1)C(=O)O

Specifications

Melting Point 30°C to 34°C
Density 1.1
Boiling Point 138°C to 140°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.499
Quantity 25 g
UN Number UN3261
Beilstein 1905520
Solubility Information Soluble in water (0.7g/L at 20°C).
Formula Weight 126.16
Percent Purity 97%
Chemical Name or Material 1-Cyclohexene-1-carboxylic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'