missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Cyclohexene-1-acetic acid, Thermo Scientific Chemicals
18.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
1-Cyclohexene-1-acetic acid used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 33°C |
Density | 1.04 g/mL |
Boiling Point | 140°C to 142°C (25 mmHg) |
Refractive Index | 1.4775 |
Quantity | 1 g |
Solubility Information | Insoluble in water. |
Formula Weight | 140.18 |
Physical Form | Liquid or Low Melting Solid |
Chemical Name or Material | 1-Cyclohexene-1-acetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'