missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexene-1-acetic acid

Product Code. 11343587
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343587 5 g 5g
11333587 1 g 1g
11353587 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343587 Supplier Thermo Scientific Alfa Aesar Supplier No. 019462.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18294-87-6 | C8H12O2 | 140.182 g/mol

1-Cyclohexene-1-acetic acid used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18294-87-6
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00015482
InChI Key KDFBPHXESBPHTK-UHFFFAOYSA-N
Synonym 1-cyclohexenylacetic acid, 1-cyclohexene-1-acetic acid, 2-cyclohex-1-en-1-yl acetic acid, cyclohex-1-enylacetic acid, 1-cyclohexene-1-aceticacid, 2-cyclohexen-1-yl acetic acid, 2-cyclohex-1-enylacetic acid, cyclohex-1-en-1-ylacetic acid, 1-cyclohexenylacetate, cyclohexenylacetic acid
PubChem CID 86699
IUPAC Name 2-(cyclohexen-1-yl)acetic acid
SMILES C1CCC(=CC1)CC(=O)O

Specifications

Melting Point 33°C
Density 1.04 g/mL
Boiling Point 140°C to 142°C (25 mmHg)
Refractive Index 1.4775
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 140.18
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 1-Cyclohexene-1-acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.