missing translation for 'onlineSavingsMsg'
Learn More

1-Cyanonaphthalene, 95%, ACROS Organics™

Product Code. 10505491 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
This item is not returnable. View return policy

Product Code. 10505491

Brand: Thermo Scientific Acros 110800025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 86-53-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 36°C to 38°C
Color Purple-Red to Red-Yellow
Boiling Point 299°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C11H7N
Linear Formula C10H7CN
MDL Number MFCD00003866
Quantity 2.5 g
Beilstein 09, 649
Synonym 1-naphthonitrile, 1-cyanonaphthalene, 1-naphthalenecarbonitrile, naphthalenecarbonitrile, 1-naphthylnitrile, alpha-naphthonitrile, 1-naphthalenenitrile, alpha-naphthylnitrile, alpha-cyanonaphthalene, unii-2as1lz61hd
InChI Key YJMNOKOLADGBKA-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC=C2C#N
IUPAC Name naphthalene-1-carbonitrile
Molecular Weight (g/mol) 153.18
PubChem CID 6846
Formula Weight 153.18
Percent Purity 95%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 1-Cyanonaphthalene
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.