missing translation for 'onlineSavingsMsg'
Learn More

1-Cyanomethyl-2-(4-cyanophenyl)-4,5-dicyanoimidazole, 97%, Thermo Scientific™

Catalog Number 15441226
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1347815-43-3
C14H6N6
258.244
MFCD20265274
OAMIMEALBFOHIE-UHFFFAOYSA-N
1-cyanomethyl-2-4-cyanophenyl-4,5-dicyanoimidazole, 1-cyanomethyl-2-4-cyanophenyl imidazole-4,5-dicarbonitrile
73995824
1-(cyanomethyl)-2-(4-cyanophenyl)imidazole-4,5-dicarbonitrile
C1=CC(=CC=C1C#N)C2=NC(=C(N2CC#N)C#N)C#N
This item is not returnable. View return policy
1347815-43-3
C14H6N6
258.244
MFCD20265274
OAMIMEALBFOHIE-UHFFFAOYSA-N
1-cyanomethyl-2-4-cyanophenyl-4,5-dicyanoimidazole, 1-cyanomethyl-2-4-cyanophenyl imidazole-4,5-dicarbonitrile
73995824
1-(cyanomethyl)-2-(4-cyanophenyl)imidazole-4,5-dicarbonitrile
C1=CC(=CC=C1C#N)C2=NC(=C(N2CC#N)C#N)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3439
Formula Weight 258.24
Percent Purity 97%
Chemical Name or Material 1-Cyanomethyl-2-(4-cyanophenyl)-4,5-dicyanoimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'