missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (1-Cyano-2-ethoxy-2-oxoethylidenaminooxy)dimethylamino-morpholinocarbenium hexafluorophosphate, 98%

Product Code. 15415175
Change view
Click to view available options
Quantity:
10g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415175 10g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15415175 Supplier Alfa Aesar™ Supplier No. H33746.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
(1-Cyano-2-ethoxy-2-oxoethylidenaminooxy)dimethylamino-morpholinocarbenium hexafluorophosphate is used as a coupling reagent in the peptide synthesis. It is suitable for solution phase & solid phase peptide synthesis, visual or colorimetric reaction monitoring possible.

Solubility
Soluble in DMF 1mmol in 2mL.

Notes
Air and moisture sensitive. Store away from air, heat, water/ moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1075198-30-9
Molecular Formula C12H19F6N4O4P
MDL Number MFCD11975052
Quantity 10g
Synonym comu, 1-cyano-2-ethoxy-2-oxoethylidenaminooxy dim, 1-cyano-2-ethoxy-2-oxoethylidenaminooxy dimethylamino-morpholinocarbenium hexafluorophosphate, 1-cyano-2-ethoxy-2-oxoethylideneaminooxy-dimethylamino-morpholino-carbenium hexafluorophosphate, z-1-cyano-2-ethoxy-2-oxoethylidene amino oxy-n,n-dimethyl morpholin-4-yl methaniminium hexafluorophosphate, 4-e-1-cyano-2-ethoxy-2-oxoethylidene amino oxidanyl dimethylamino methylidene-4??-morpholin-4-ylium hexafluorophosphate
InChI Key GPDHNZNLPKYHCN-DZOOLQPHSA-N
SMILES CCOC(=O)C(=NOC(=[N+](C)C)N1CCOCC1)C#N.F[P-](F)(F)(F)(F)F
IUPAC Name [[(Z)-(1-cyano-2-ethoxy-2-oxoethylidene)amino]oxy-morpholin-4-ylmethylidene]-dimethylazanium;hexafluorophosphate
Molecular Weight (g/mol) 428.272
PubChem CID 44471148
Formula Weight 428.27
Percent Purity 98%
Chemical Name or Material (1-Cyano-2-ethoxy-2-oxoethylidenaminooxy)dimethylamino-morpholinocarbenium hexafluorophosphate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.