missing translation for 'onlineSavingsMsg'
Learn More

1-Chloropropane, 99%

Product Code. 10259223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10259223 500 mL Glass bottle 500mL
10244470 5 mL Glass bottle 5mL
10794211 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10259223 Supplier Thermo Scientific Acros Supplier No. 109955000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 540-54-5 | C3H7Cl | 78.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-54-5
Molecular Formula C3H7Cl
Molecular Weight (g/mol) 78.54
MDL Number MFCD00000995
InChI Key SNMVRZFUUCLYTO-UHFFFAOYSA-N
Synonym propyl chloride, propane, 1-chloro, n-propyl chloride, chloropropane, unii-tuv7462nwk, 1-chloro-propane, tuv7462nwk, n-propylchloride, propane, chloro, 1-chloropropyl
PubChem CID 10899
IUPAC Name 1-chloropropane
SMILES CCCCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -123°C
Color Colorless
Density 0.8920g/mL
Boiling Point 46°C to 47°C
Flash Point -18°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2Cl
Refractive Index 1.3870 to 1.389
Quantity 500 mL
Beilstein 01, 104
Merck Index 15, 7960
Specific Gravity 0.892
Solubility Information Solubility in water: 2.7g/L (20°C). Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 78.54
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-chloropropane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chloropropane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.