missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorophthalazin-4-one, 98%

Product Code. p-7029990
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15433707

Brand: Thermo Scientific Alfa Aesar H27515.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2257-69-4 | C8H5ClN2O | 180.59 g/mol

1-Chlorophthalazin-4-one acts as an organic and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorophthalazin-4-one acts as an organic and pharmaceutical intermediate.

Notes
Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2257-69-4
Molecular Formula C8H5ClN2O
Molecular Weight (g/mol) 180.59
MDL Number MFCD00102192
InChI Key QCKGMJDOJRNSMS-UHFFFAOYSA-N
Synonym 4-chlorophthalazin-1 2h-one, 4-chloro-1,2-dihydrophthalazin-1-one, 4-chlorophthalazin-1-ol, 1 2h-phthalazinone, 4-chloro, 4-chloro-1 2h-phthalazinone, 4-chlorophthalazone, 4-chloro-1-phthalazinol, 4-chlorophthalazin-1-one, 1-chlorophthalazin-4-one, 1-chlorophthalazine-4-one
PubChem CID 2775142
SMILES ClC1=NNC(=O)C2=CC=CC=C12

Specifications

Melting Point 270°C to 274°C
Quantity 250 mg
IUPAC Name 4-chloro-2H-phthalazin-1-one
Formula Weight 180.59
Percent Purity 98%
Chemical Name or Material 1-Chlorophthalazin-4-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.