missing translation for 'onlineSavingsMsg'
Learn More

1-Chloropentane, TRC

Product Code. 30361665
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30361665 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30361665

Brand: Toronto Research Chemicals TRCC3737805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Chloropentane
CAS 543-59-9
Molecular Formula C5H11Cl
Quantity 5 g
Synonym Amyl Chloride, NSC 7898, Pentyl Chloride, n-Amyl Chloride, n-Pentyl Chloride
InChI Formula InChI=1S/C5H11Cl/c1-2-3-4-5-6/h2-5H2,1H3
SMILES CCCCCCl
IUPAC Name 1-chloropentane
Molecular Weight (g/mol) 106.59
Formula Weight 106.0549
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.