missing translation for 'onlineSavingsMsg'
Learn More

1-Chloropentane, 98%

Product Code. 11446426
Change view
Click to view available options
Quantity:
1000 g
Unit Size:
1000g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446426 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11446426 Supplier Thermo Scientific Alfa Aesar Supplier No. B24733.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 543-59-9 | C5H11Cl | 106.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-59-9
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.59
MDL Number MFCD00001015
InChI Key SQCZQTSHSZLZIQ-UHFFFAOYSA-N
Synonym pentyl chloride, pentane, 1-chloro, n-amyl chloride, n-pentyl chloride, amyl chloride, chloropentane, unii-0eg9msd3nk, 0eg9msd3nk, pentane, chloro, n-butylcarbonyl chloride
PubChem CID 10977
IUPAC Name 1-chloropentane
SMILES CCCCCCl

Specifications

Melting Point -99°C
Density 0.881
Boiling Point 106°C to 108°C
Flash Point 3°C (37°F)
Odor Sweet
Linear Formula CH3(CH2)4Cl
Refractive Index 1.4125
Quantity 1000 g
UN Number UN1107
Merck Index 14,606
Formula Weight 106.6
Percent Purity 98%
Chemical Name or Material 1-Chloropentane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.