missing translation for 'onlineSavingsMsg'
Learn More

1-Chloronaphthalene (Technical Grade), TRC

Product Code. 30362448
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
Product Code. Quantity unitSize
30362448 250 g 250g
1 options
This item is not returnable. View return policy

Product Code. 30362448

Brand: Toronto Research Chemicals TRCC370910250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Chloronaphthalene
CAS 90-13-1
Molecular Formula C10 H7 Cl
Quantity 250 g
Synonym Naphthalene, 1-chloro-, 1-Chloronaphthalene, 1-Chloronaphthaline, 1-Naphthalenyl chloride, 1-Naphthyl chloride, NSC 6166, PCN 1, α-Chloronaphthalene, α-Naphthyl chloride, 1-Monochloronaphthalene, 1-MonoCN, 1-MCN, PCN-1
InChI Formula InChI=1S/C10H7Cl/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H
SMILES Clc1cccc2ccccc12
IUPAC Name 1-chloronaphthalene
Molecular Weight (g/mol) 162.62
Formula Weight 162.0236
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.