missing translation for 'onlineSavingsMsg'
Learn More

1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene

Product Code. 10575671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10575671 1 L Glass bottle 1 L
10479252 100 mL Glass bottle 100mL
11954521 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10575671 Supplier Thermo Scientific Acros Supplier No. 180970010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-13-1 | C10H7Cl | 162.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-13-1 , 91-58-7
Molecular Formula C10H7Cl
Molecular Weight (g/mol) 162.62
InChI Key JTPNRXUCIXHOKM-UHFFFAOYSA-N
Synonym alpha-chloronaphthalene, naphthalene, 1-chloro, 1-naphthyl chloride, chloronaphthalene, 1-chloro-naphthalene, alpha-naphthyl chloride, 1-chlornaftalen, 1-chloro naphthalene, alpha-chlornaphthalene, .alpha.-chloronaphthalene
PubChem CID 7003
IUPAC Name 1-chloronaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Cl

Specifications

Melting Point -2.5°C
Color Colorless to Yellow
Density 1.1940g/mL
Boiling Point 259.0°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (HPLC)
Packaging Glass bottle
Linear Formula C10H7Cl
Refractive Index 1.6310 to 1.6340
Quantity 1 L
Beilstein 05,541
Merck Index 15,2151
Specific Gravity 1.194
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene,petroleum,ether and alcohol
Formula Weight 162.62
Percent Purity 85%
Grade Technical
Physical Form Liquid
Chemical Name or Material 1-Chloronaphthalene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chloronaphthalene, ca. 85-90%, remainder 2-Chloronaphthalene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.