missing translation for 'onlineSavingsMsg'
Learn More

1-(Chloromethyl)naphthalene, 95%

Product Code. p-88356 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10365930

Brand: Thermo Scientific 109450050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-52-2 | C11H9Cl | 176.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-52-2
Molecular Formula C11H9Cl
Molecular Weight (g/mol) 176.65
MDL Number MFCD00004042
InChI Key XMWGTKZEDLCVIG-UHFFFAOYSA-N
Synonym 1-chloromethyl naphthalene, naphthalene, 1-chloromethyl, 1-naphthylmethyl chloride, 1-menaphthyl chloride, 1-chlormethyl naftalen, chloromethyl naphthalene, 1-chloro methylnaphthalene, alpha-naphthylmethyl chloride
PubChem CID 6845
IUPAC Name 1-(chloromethyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2CCl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 31°C to 31°C
Color Yellow to Brown
Density 1.1700g/mL
Boiling Point 291°C to 292°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range Remainder 2-chloromethylnaphtalene (HPLC)
Packaging Glass bottle
Linear Formula C10H7CH2Cl
Refractive Index 1.6350 to 1.639
Quantity 5 g
Beilstein 05, 566
Specific Gravity 1.17
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 176.65
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material 1-(Chloromethyl)naphthalene, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(Chloromethyl)naphthalene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.